CAS RN 16600-92-3
IUPAC Navn:
2-nitrobenzene-1,3,5-triol
Synonymer:
2-nitrophloroglucinol
Molekylvægt (g/mol):
171.11
Molekylformel:
C6H5NO5
InChi Key:
QSVQZFVXAUGEMT-UHFFFAOYSA-N
SMILES:
OC1=CC(O)=C(C(O)=C1)[N+]([O-])=O
1
–
1
af
1
Resultater
1
2-Nitrophloroglucinol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.