CAS RN 165739-64-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-Chloro Loratadine, TRC
CAS: 165739-64-0 Molekylær formel: C22 H22 Cl2 N2 O2 Molekylvægt (g/mol): 417.33 Synonym: Ethyl 4-(2,8-Dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate IUPAC navn: ethyl 4-(2,8-dichloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-c]pyridin-11-ylidene)piperidine-1-carboxylate SMIL: CCOC(=O)N1CCC(=C2c3ccc(Cl)cc3CCc4ccc(Cl)nc24)CC1
2
Ethyl 4-(2,8-Dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.