CAS RN 16503-22-3
IUPAC Navn:
[2-(1H-imidazol-5-yl)ethyl](methyl)amine dihydrochloride
Synonymer:
[2-(3H-imidazol-4-yl)ethyl](methyl)amine dihydrochloride
Molekylvægt (g/mol):
198.09
Molekylformel:
C6H13Cl2N3
InChi Key:
AYUQICXJAMPXPF-UHFFFAOYSA-N
SMILES:
Cl.Cl.CNCCC1=CN=CN1
1
–
1
af
1
Resultater
1
N-a-Methylhistamine Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.