CAS RN 1610-18-0
IUPAC Navn:
6-methoxy-N2,N4-bis(propan-2-yl)-1,3,5-triazine-2,4-diamine
Synonymer:
ontrack methoxypropazine prometone pramitol gesafram prometon
Molekylvægt (g/mol):
225.3
Molekylformel:
C10H19N5O
InChi Key:
ISEUFVQQFVOBCY-UHFFFAOYSA-N
SMILES:
COC1=NC(NC(C)C)=NC(NC(C)C)=N1
1
–
7
af
7
Resultater
2
Prometon, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Prometon, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.