CAS RN 154754-57-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
6-(8-Chloro-4,5-dihydro-1-methyl-6H-imidazo[1,5-a][1,4]benzodiazepin-6-ylidene)-2,4-cyclohexadien-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-(8-Chloro-4,5-dihydro-1-methyl-6H-imidazo[1,5-a][1,4]benzodiazepin-6-ylidene)-2,4-cyclohexadien-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.