CAS RN 1542213-00-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
NCT-502, MedChemExpress
MedChemExpress NCT-502 is a human phosphoglycerate dehydrogenase (PHGDH) inhibitor, cytotoxic to PHGDH-dependent cancer cells, and reduces glucose-derived serine production, with an IC50 of 3.7 μM against PHGDH.