CAS RN 152-43-2
IUPAC Navn:
(1R,3aS,3bR,9bS,11aS)-7-(cyclopentyloxy)-1-ethynyl-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-ol
Synonymer:
quinestrol quinestrol (product) plestrovis estrovis ethinyl estradiol 3 cyclopentyl ether qui-lea quinestrolo eston quilea estrovister
Molekylvægt (g/mol):
364.53
Molekylformel:
C25H32O2
InChi Key:
PWZUUYSISTUNDW-VAFBSOEGSA-N
SMILES:
C[C@]12CC[C@H]3[C@@H](CCC4=CC(OC5CCCC5)=CC=C34)[C@@H]1CC[C@@]2(O)C#C
1
–
1
af
1
Resultater
1
Quinestrol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.