CAS RN 150375-75-0
IUPAC Navn:
(2S)-1-[(2R,3S)-5-chloro-3-(2-chlorophenyl)-1-(3,4-dimethoxybenzenesulfonyl)-3-hydroxy-2,3-dihydro-1H-indole-2-carbonyl]pyrrolidine-2-carboxamide
Synonymer:
relcovaptan
Molekylvægt (g/mol):
620.5
Molekylformel:
C28H27Cl2N3O7S
InChi Key:
CEBYCSRFKCEUSW-NAYZPBBASA-N
SMILES:
COC1=CC=C(C=C1OC)S(=O)(=O)N1[C@@H](C(=O)N2CCC[C@H]2C(N)=O)[C@](O)(C2=CC(Cl)=CC=C12)C1=CC=CC=C1Cl
1
–
4
af
4
Resultater
1
Relcovaptan, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tocris Bioscience™ SR 49059
Selective, orally active vasopressin V1A receptor antagonist
3
Relcovaptan, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Relcovaptan, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.