CAS RN 1497415-41-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ SB 24011
Inhibitor of STING-TRIM29 interaction; increases cellular STING levels