CAS RN 149198-47-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(3R,4S)-1-t-Boc-3-[(triethylsilyl)oxy]-4-phenyl-2-azetidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3R,4S)-1-t-Boc-3-[(triethylsilyl)oxy]-4-phenyl-2-azetidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3R,4S)-1-t-Boc-3-[(triethylsilyl)oxy]-4-phenyl-2-azetidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.