CAS RN 148218-14-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
14-Bromo, 13-Deoxo, 13,13-Dimethoxy Daunorubicin Hydrobromide (>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(8S,10S)-10-[(3-Amino-2,3,6-trideoxy-Alpha-L-lyxo-hexopyranosyl)oxy]-8-(2-bromo-1,1-dimethoxyethyl)-7,8,9,10-tetrahydro-6,8,11-trihydroxy-1-methoxy-5,12-naphthacenedione Hydrobromide(>85%)(Doxorubicin Impurity) Hydrobromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.