CAS RN 147676-94-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
1,4-Dihydro-5-[5-(2-bromoacetyl)-2-ethoxyphenyl]-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one (may contain 5-(2-chloroacetyl)), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,4-Dihydro-5-[5-(2-bromoacetyl)-2-ethoxyphenyl]-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one (may contain 5-(2-chloroacetyl)), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.