CAS RN 1470374-22-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
MedChemExpress TAK-448 acetate
MedChemExpress TAK-448 acetate (MVT-602 acetate) is a potent and full KISS1R agonist with an IC50 of 460 pM and an EC50 of 632 pM.
2
MedChemExpress TAK-448 acetate
MedChemExpress TAK-448 acetate (MVT-602 acetate) is a potent and full KISS1R agonist with an IC50 of 460 pM and an EC50 of 632 pM.