CAS RN 145783-15-9
IUPAC Navn:
4,6-dichloro-2-(propylsulfanyl)pyrimidin-5-amine
Synonymer:
4,6-dichloro-2-(propylsulfanyl)pyrimidin-5-amine
Molekylvægt (g/mol):
238.13
Molekylformel:
C7H9Cl2N3S
InChi Key:
CJJLJBFJNXMANZ-UHFFFAOYSA-N
SMILES:
CCCSC1=NC(Cl)=C(N)C(Cl)=N1
1
–
1
af
1
Resultater
1
4,6-Dichloro-2-(propylthio)pyrimidin-5-amine, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.