CAS RN 14545-23-4
IUPAC Navn:
(²H₅)aniline
Synonymer:
(²H₅)aniline
Molekylvægt (g/mol):
100.17
Molekylformel:
C6H7N
InChi Key:
PAYRUJLWNCNPSJ-LQHBDRBESA-N
SMILES:
[2H]N([2H])C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H]
1
–
1
af
1
Resultater
1
Aniline-d7, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.