CAS RN 1448460-32-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
17-alpha-Hydroxymethyl-17-beta-methyl-18-nor-2-oxa-5-alpha-androst-13-en-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
17alpha-Hydroxymethyl-17beta-methyl-18-nor-2-oxa-5alpha-androst-13-en-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
17alpha-Hydroxymethyl-17Beta-methyl-18-nor-2-oxa-5alpha-androst-13-en-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.