CAS RN 14484-64-1
IUPAC Navn:
iron(3+) tris((dimethylcarbamothioyl)sulfanide)
Synonymer:
ferbam
Molekylvægt (g/mol):
416.47
Molekylformel:
C9H18FeN3S6
InChi Key:
WHDGWKAJBYRJJL-UHFFFAOYSA-K
SMILES:
[Fe+3].CN(C)C([S-])=S.CN(C)C([S-])=S.CN(C)C([S-])=S
1
–
2
af
2
Resultater
1
Ferbam, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Ferbam, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.