CAS RN 1446182-94-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
CUDC-427, MedChemExpress
MedChemExpress CUDC-427 is a potent second-generation pan-selective IAP antagonist, used for treatment of various cancers.