CAS RN 1428728-83-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2,2'-((6,6'-(2,4-Di(pyridin-2-yl)cyclobutane-1,3-diyl)bis(1H-indazole-6,3-diyl))bis(sulfanediyl))bis(N-methylbenzamide), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,2'-((6,6'-(2,4-Di(pyridin-2-yl)cyclobutane-1,3-diyl)bis(1H-indazole-6,3-diyl))bis(sulfanediyl))bis(N-methylbenzamide), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.