CAS RN 14277-97-5
IUPAC Navn:
trisodium (2S,3S,4S)-4-[(2Z,4E,6R)-6-carboxylato-6-methylhexa-2,4-dien-2-yl]-3-(carboxylatomethyl)pyrrolidine-2-carboxylate
Synonymer:
domoic acid
Molekylvægt (g/mol):
377.28
Molekylformel:
C15H18NNa3O6
InChi Key:
DKZXEHCSMAYDTA-LSCPQTDESA-K
SMILES:
[Na+].[Na+].[Na+].C[C@H](\C=C\C=C(\C)[C@H]1CN[C@@H]([C@H]1CC([O-])=O)C([O-])=O)C([O-])=O
1
–
1
af
1
Resultater
1
Domoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.