CAS RN 1425043-73-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
IPI-3063, MedChemExpress
MedChemExpress IPI-3063 is a potent and selective PI3K p110δ inhibitor with an IC50 of 2.5 ± 1.2 nM.