CAS RN 14215-97-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
1-O-Acetyl-2,3,5-tri-O-benzoyl-Alpha,Beta-D-ribofuranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-O-Acetyl-2,3,5-tri-O-benzoyl-Alpha,Beta-D-ribofuranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-O-Acetyl-2,3,5-tri-O-benzoyl-Alpha,Beta-D-ribofuranose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.