CAS RN 1415238-77-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
5S rRNA modificator, MedChemExpress
MedChemExpress 5S rRNA modificator is a suitable electrophile for 2’-hydroxyl acylation on structured RNA molecules, yielding accurate structural information comparable to that obtained with existing probes; 5S rRNA RNA modification.
2
Tocris Bioscience™ FAI
Reagent for RNA SHAPE-MaP experiments in live cells