CAS RN 1401728-56-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
NE 52-QQ57, MedChemExpress
MedChemExpress NE 52-QQ57 is a selective, and orally available GPR4 antagonist with an IC50 of 70 nM. NE 52-QQ57 has anti-inflammatory activity.