CAS RN 1400690-70-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Kampagner er tilgængelige
2-(N-Methylperfluorooctanesulfonamido)acetic acid D3 (N-methyl D3) 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
2-(N-Methylperfluorooctanesulfonamido)acetic acid D3 (N-methyl D3) 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Kampagner er tilgængelige
Dr. Ehrenstorfer Mass-Labelled PFAS-24ES Extraction Mixture 1 μg/mL in Methanol:Water
Dr. Ehrenstorfer Mass-Labelled PFAS-24ES Extraction Mixture 1 μg/mL in Methanol:Water
Certified reference materials available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality.