CAS RN 1394013-66-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
3,6,9,16,19,22-Hexaoxabicyclo[22.3.1]octacosa-1(28),24,26-triene-2,10,15,23-tetrone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,6,9,16,19,22-Hexaoxabicyclo[22.3.1]octacosa-1(28),24,26-triene-2,10,15,23-tetrone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3,6,9,16,19,22-Hexaoxabicyclo[22.3.1]octacosa-1(28),24,26-triene-2,10,15,23-tetrone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.