CAS RN 139059-00-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(4R-5R)-4,5-Dihydro-5-[4-(methylsulfonyl)phenyl]-2-phenyl-4-oxazolecarboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(4R-5R)-4,5-Dihydro-5-[4-(methylsulfonyl)phenyl]-2-phenyl-4-oxazolecarboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(4R-5R)-4,5-Dihydro-5-[4-(methylsulfonyl)phenyl]-2-phenyl-4-oxazolecarboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.