CAS RN 1379779-17-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2,5-Dihydro-3-(hydroxymethyl)-2,2,5,5-tetramethyl-4-(3-pyridinyl)-1H-pyrrol-1-yloxy, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,5-Dihydro-3-(hydroxymethyl)-2,2,5,5-tetramethyl-4-(3-pyridinyl)-1H-pyrrol-1-yloxy, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.