CAS RN 1373919-90-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ IRDye™ 680RD, Maleimide
Near-infared dye (reactive group maleimide)