CAS RN 1372695-75-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(3alpha,17alpha)-4-Chloro-3-hydroxy-17-methyl-18-Norandrosta-4,13-diene-17-methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.