CAS RN 136565-73-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Anemarsaponin E, MedChemExpress
MedChemExpress Anemarsaponin E is extracted from Anemarrhena asphodeloides Bunge and has anti-inflammatory activity.