CAS RN 13649-84-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(16Beta)-21-(Acetyloxy)-2'-methyl-5'H-Pregna-1,4-dieno[17,16-d]oxazole-3,11,20-trione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(16b)-21-(Acetyloxy)-2'-methyl-5'H-Pregna-1,4-dieno[17,16-d]oxazole-3,11,20-trione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(16Beta)-21-(Acetyloxy)-2'-methyl-5'H-Pregna-1,4-dieno[17,16-d]oxazole-3,11,20-trione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.