CAS RN 1347744-96-0
IUPAC Navn:
N-(3-methylphenyl)-2-[4-(pyridin-2-yl)piperidin-1-yl]acetamide dihydrochloride
Synonymer:
N-(3-methylphenyl)-2-[4-(pyridin-2-yl)piperidin-1-yl]acetamide dihydrochloride
Molekylvægt (g/mol):
382.33
Molekylformel:
C19H25Cl2N3O
InChi Key:
KIGNSSHMCNBHQX-UHFFFAOYSA-N
SMILES:
Cl.Cl.CC1=CC(NC(=O)CN2CCC(CC2)C2=CC=CC=N2)=CC=C1