CAS RN 132869-83-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
MedChemExpress HOE-S 785026
MedChemExpress HOE-S 785026 is a blue fluorescent dyes, which can be used as a cell dye for DNA.