CAS RN 132014-87-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
alpha-Methyl-Gamma-hydroxy-1,N2-propano-2’-deoxyguanosine (Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
a-Methyl-gamma-hydroxy-1,N2-propano-2’-deoxyguanosine(Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.