CAS RN 1311982-88-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
MIR96-IN-1, MedChemExpress
MedChemExpress MIR96-IN-1 targets the Drosha site in the miR-96 (miRNA-96, microRNA-96) hairpin precursor, inhibiting its biogenesis, derepressing downstream targets, and triggering apoptosis in breast cancer cells. MIR96-IN-1 binds to RNAs with Kds of 1.3, 9.4, 3.4, 1.3 and 7.4 μM for RNA1, RNA2, RNA3, RNA4 and RNA5, respectively.