CAS RN 130914-77-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
[2-[(2-Amino-6-oxo-3H-purin-9-yl)methoxy]-3-[(2S)-2-(benzyloxycarbonylamino)-3-methyl-butanoyl]oxy-propyl] (2S)-2-(Benzyloxycarbonylamino)-3-methylbutanoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.