CAS RN 13049-40-6
IUPAC Navn:
4,4'-diethyl-1,1'-biphenyl
Synonymer:
4,4'-diethyl-1,1'-biphenyl
Molekylvægt (g/mol):
210.32
Molekylformel:
C16H18
InChi Key:
UMSGIWAAMHRVQI-UHFFFAOYSA-N
SMILES:
CCC1=CC=C(C=C1)C1=CC=C(CC)C=C1
1
–
3
af
3
Resultater
1
4,4'-Diethylbiphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4,4'-Diethylbiphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4,4'-Diethylbiphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.