CAS RN 128055-74-3
IUPAC Navn:
2,2',7,7'-tetrabromo-9,9'-spirobi[fluorene]
Synonymer:
2,2',7,7'-tetrabromo-9,9'-spirobi[fluorene]
Molekylvægt (g/mol):
631.99
Molekylformel:
C25H12Br4
InChi Key:
MASXXNUEJVMYML-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)C21C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
1
–
2
af
2
Resultater
1
2,2',7,7'-Tetrabromo-9,9'-spirobifluorene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,2',7,7'-Tetrabromo-9,9'-spirobifluorene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.