CAS RN 127516-23-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Impurity of Doxercalciferol, MedChemExpress
MedChemExpress Impurity of Doxercalciferol is an impurity of doxercalciferol, which is a synthetic analog of ergocalciferol (vitamin D2), used as a drug for secondary hyperparathyroidism and metabolic bone disease, and it suppresses parathyroid synthesis and secretion.