CAS RN 127243-85-0
IUPAC Navn:
N-(2-{[(2E)-3-(4-bromophenyl)prop-2-en-1-yl]amino}ethyl)isoquinoline-5-sulfonamide
Synonymer:
N-(2-{[(2E)-3-(4-bromophenyl)prop-2-en-1-yl]amino}ethyl)isoquinoline-5-sulfonamide
Molekylvægt (g/mol):
446.36
Molekylformel:
C20H20BrN3O2S
InChi Key:
ZKZXNDJNWUTGDK-NSCUHMNNSA-N
SMILES:
BrC1=CC=C(\C=C\CNCCNS(=O)(=O)C2=C3C=CN=CC3=CC=C2)C=C1
1
–
3
af
3
Resultater
1
N-[2-(p-Bromocinnamylamino)ethyl]-5-Isoquinoline Sulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[2-(p-Bromocinnamylamino)ethyl]-5-Isoquinoline Sulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-[2-(p-Bromocinnamylamino)ethyl]-5-Isoquinoline Sulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.