CAS RN 1269489-98-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
2-(Acetylamino)-2-deoxy-N-methoxy-N-methyl-3,4,6-tris-O-(phenylmethyl)-Beta-D-glucopyranosylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(Acetylamino)-2-deoxy-N-methoxy-N-methyl-3,4,6-tris-O-(phenylmethyl)-b-D-glucopyranosylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(Acetylamino)-2-deoxy-N-methoxy-N-methyl-3,4,6-tris-O-(phenylmethyl)-Beta-D-glucopyranosylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.