CAS RN 1267610-30-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Tocris Bioscience™ LDN 193188
Inhibitor of phosphatidylcholine transfer protein (PC-TP)
2
3