CAS RN 1260590-51-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
N-Methyl-N-[(3R,4R)-4-methyl-3-piperidinyl]-7H-Pyrrolo[2,3-d]pyrimidin-4-amine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Methyl-N-[(3R,4R)-4-methyl-3-piperidinyl]-7H-Pyrrolo[2,3-d]pyrimidin-4-amine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Methyl-N-[(3R,4R)-4-methyl-3-piperidinyl]-7H-Pyrrolo[2,3-d]pyrimidin-4-amine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.