CAS RN 125112-76-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(3Alpha,5Beta,6Alpha,7Alpha)-6,7-Epoxy-3-[(ethoxycarbonyl)oxy]cholan-24-oic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3Alpha,5Beta,6Alpha,7Alpha)-6,7-Epoxy-3-[(ethoxycarbonyl)oxy]cholan-24-oic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.