CAS RN 1244762-86-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(2'R)-2'-Deoxy-3'-O-[(1,1-dimethylethyl)dimethylsilyl]-2'-fluoro-2'-methyl-uridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2'R)-2'-Deoxy-3'-O-[(1,1-dimethylethyl)dimethylsilyl]-2'-fluoro-2'-methyl-uridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2'R)-2'-Deoxy-3'-O-[(1,1-dimethylethyl)dimethylsilyl]-2'-fluoro-2'-methyl-uridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.