CAS RN 1235828-02-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(S)-1-(Benzyloxy)-3-(3,4-bis((tert-butyldimethylsilyl)oxy)phenyl)propan-2-yl acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(S)-1-(Benzyloxy)-3-(3,4-bis((tert-butyldimethylsilyl)oxy)phenyl)propan-2-yl acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(S)-1-(Benzyloxy)-3-(3,4-bis((tert-butyldimethylsilyl)oxy)phenyl)propan-2-yl acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.