CAS RN 1220393-12-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(3R)-3-[[2-(1,3-Benzodioxol-5-ylcarbonyl)-1H-indol-3-yl]methyl]-1-methyl-2,5-piperazinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3R)-3-[[2-(1,3-Benzodioxol-5-ylcarbonyl)-1H-indol-3-yl]methyl]-1-methyl-2,5-piperazinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.