CAS RN 1206164-60-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(5Alpha,6Alpha,17Beta)-6,17-Dihydroxy-17-methylandrostan-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(5Alpha,6Alpha,17Beta)-6,17-Dihydroxy-17-methylandrostan-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(5a,6a,17b)-6,17-Dihydroxy-17-methylandrostan-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.