CAS RN 1189419-70-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
2-(tert-Butylamino)-4-chloro-6-cyclopropylamino-1,3,5-triazine-d9, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-tert-Butylamino-d9-4-chloro-6-cyclopropylamino-1,3,5-triazine, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
3
2-(tert-Butylamino)-4-chloro-6-cyclopropylamino-1,3,5-triazine-d9, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.